Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

VLDLR Rabbit anti-Human, Rat, Clone: VLDLR/2896R, Novus Biologicals™

Rabbit Monoclonal Antibody

$159.82 - $406.52

Specifications

Antigen VLDLR
Clone VLDLR/2896R
Concentration 0.2 mg/ml
Dilution ELISA
Applications ELISA
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
NB121869
View Documents
Novus Biologicals
NBP30719810UG
100 μg
Each for $406.52
Add to cart
 
NB121870
View Documents
Novus Biologicals
NBP30719820UG
20 μg
Each for $159.82
Add to cart
 
Description

Description

VLDLR Monoclonal antibody specifically detects VLDLR in Human, Rat samples. It is validated for ELISA
Specifications

Specifications

VLDLR
0.2 mg/ml
ELISA
Purified
RUO
Human, Rat
CARMQ1, CHRMQ1, FLJ35024, very low density lipoprotein receptor, very low-density lipoprotein receptor, VLDL receptor, VLDL-R, VLDLRCH
Recombinant human VLDLR fragment from c-terminal
Monoclonal
Protein A or G purified
VLDLR/2896R
ELISA
Unconjugated
Rabbit
Apoptosis, Cholesterol Metabolism, Lipid and Metabolism, Neurodegeneration, Neuronal Cell Markers, Neuroscience
10 mM PBS with 0.05% BSA
7436
IgG
Primary
Store at 4°C.
SDS
Product Certifications
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit