Learn More
Waters Corp High Pressure Motorized Valve Rebuild Kit
Supplier:  Waters Corp WAT045424
Encompass
The high pressure valve rebuild kit contains all of the Waters Quality Parts™ required to fully rebuild the high pressure valve of the Waters 717 Plus Autosampler, 2695 Separations Modules and 2695D Separations Modules. The spool is crafted to precise hardness, smoothness, and alignment specifications. The high-pressure valve seals are made of proprietary materials and then packed in anti-static bags to further minimize the chance of particulate contamination. The rebuild kit contains a PTFE O-ring, seal, spool andhigh pressure seals.
Catalog No. NC1335912