Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

VMAT2 Mouse anti-Human, Clone: OTI10C11, Invitrogen™

Mouse Monoclonal Antibody

Manufacturer:  Invitrogen MA524930

Catalog No. PIMA524930


Add to cart

Description

Description

The vesicular monoamine transporter acts to accumulate cytosolic monoamines into synaptic vesicles, using the proton gradient maintained across the synaptic vesicular membrane. Its proper function is essential to the correct activity of the monoaminergic systems that have been implicated in several human neuropsychiatric disorders. The transporter is a site of action of important drugs, including reserpine and tetrabenazine.
Specifications

Specifications

VMAT2
Monoclonal
1.0 mg/mL
PBS with 1% BSA, 50% glycerol and 0.02% sodium azide; pH 7.3
Q05940
SVAT, SVMT, VAT2, VMAT2, monoamine neurotransmitter transporter, monoamine transporter, solute carrier family 18 member 2, synaptic vesicular amine transporter, vesicle monoamine transporter type 2, vesicle monoamine/H+ antiporter, vesicular amine transporter 2
Mouse
IgG1
100 μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
6571
Immunohistochemistry (Paraffin)
OTI10C11
Unconjugated
SLC18A2
Liquid
SLC18A2
Full-length protein expressed in 293T cell transfected with human SLC18A2 expression vector
Affinity Chromatography
RUO
Antibody
Monoclonal
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit