Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Bestrophin 3 Mouse anti-Human, Clone: OTI2H3, Invitrogen™

Mouse Monoclonal Antibody

Manufacturer:  Invitrogen MA525306

Catalog No. PIMA525306


Add to cart

Description

Description

BEST3 belongs to the bestrophin family of anion channels, which includes BEST1 (MIM 607854), the gene mutant in vitelliform macular dystrophy (VMD; MIM 153700), and 2 other BEST1-like genes, BEST2 (MIM 607335) and BEST4 (MIM 607336). Bestrophins are transmembrane (TM) proteins that share a homology region containing a high content of aromatic residues, including an invariant arg-phe-pro (RFP) motif. The bestrophin genes share a conserved gene structure, with almost identical sizes of the 8 RFP-TM domain-encoding exons and highly conserved exon-intron boundaries. Each of the 4 bestrophin genes has a unique 3-prime end of variable length.
Specifications

Specifications

Bestrophin 3
Monoclonal
0.98 mg/mL
PBS with 1% BSA, 50% glycerol and 0.02% sodium azide; pH 7.3
Q8N1M1
BEST3, Bestrophin-3, VMD2L3, Bestrophin 3
Mouse
IgG1
100 μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
144453
Flow Cytometry, Immunocytochemistry, Immunofluorescence, Western Blot
OTI2H3
Unconjugated
BEST3
Liquid
BEST3
Full length human recombit protein of BEST3 produced in HEK293T cell
Affinity Chromatography
RUO
Antibody
Monoclonal
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit