Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

VN1R5 Rabbit anti-Human, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA549565

Catalog No. PA549565

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Description

Description

Putative pheromone receptor.
Specifications

Specifications

VN1R5
Polyclonal
PBS with 150mM NaCl, 50% glycerol and 0.02% sodium azide; pH 7.4
Q7Z5H4
Vomeronasal type-1 receptor 5, G-protein coupled receptor GPCR26, hGPCR26, V1r-like receptor 5, VN1R5, V1RL5
Rabbit
IgG
100 μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
317705
Immunocytochemistry, Immunofluorescence, Western Blot
Unconjugated
VN1R5
Liquid
VN1R5
synthetic peptide corresponding to 62-90aa of human VN1R5
Affinity chromatography
RUO
Antibody
Polyclonal
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit