Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tocris™ Voglibose

Orally active alpha-glucosidase inhibitor

Manufacturer:  Tocris™ 3596/50

 View more versions of this product

Catalog No. 35-965-0R


  Check Availability
Add to cart

Disclaimers:

For Research Use Only


Specifications

Description

Orally active α-glucosidase inhibitor (IC50 values are 3.9 and 6.4nM at sucrase and maltase respectively). Increases glucagon-like peptide 1 (GLP-1) secretion and decreases food consumption in ob/ob mice, and reduces plasma concentrations of glucose, triglycerides and insulin in Wistar fatty rats. Exhibits antidiabetic and antiobesity activity in vivo.
  • Chemical Name: 3,4-Dideoxy-4-[[2-hydroxy-1-(hydroxymethyl)ethyl]amino]-2-C-(hydroxymethyl)-D-epi-inositol
  • Formula: C10H21NO7
  • CAS Number: 83480-29-9
  • Solubility: Soluble to 100mM in water and to 75mM in DMSO
Description & Specifications

Specifications

Voglibose
Voglibose
267.28
50mg
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit