Learn More
Volatile GC/MS Calibration Standard (BFB) (For EPA 524.2, 624, 8240, 8260, CLP)
4-Bromofluorobenzene at 2000µg/mL in methanol
$51.15
Specifications
Chemical Name or Material | Volatile GC/MS Calibration Standard (BFB) |
---|---|
Alpha Vector | Bromofluorobenzene |
Methods | For EPA Methods 524.2, 624, 8240 |
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity & Availability | ||||
US-STS-110N-1
![]() |
Agilent Technologies
STS110N1 |
1 x 1mL | Ampule |
Each for $51.15
|
|
Specifications
Volatile GC/MS Calibration Standard (BFB) | |
Bromofluorobenzene | |
For EPA Methods 524.2, 624, 8240 |