Learn More
Walter Products™ Voltage and Electric Current Relational Experiment
Shop All Walter Products Products

Description
- All experiment parts are installed within the container
- Great for safe keeping and easy transportation
Specifications
Specifications
| Grade | 5+ |
| Includes | Square Galvanometer, Ammeter, 3 Battery Compartments, 14 Terminals, Switch, 4 (10 Ω) Resistors, 3 AA Batteries, Conducting Wires with Plugs |
| Class Size | 1 Student |
| Product Type | Classroom Experiment |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.