Learn More
Vomeronasal 1 Receptor B2/B4 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Supplier:  Thermo Scientific OSR00235W
Catalog No. PIOSR00235W
Item Discontinued This item has been discontinued by the supplier and is no longer available. Please reach out to your sales representative.
Description
Description
Reconstitute with 100 μL of distilled water. Centrifuge to remove any insoluble material.
Putative pheromone receptor implicated in the regulation of social and reproductive behavior. Expressed in a subset of sensory neurons located in the apical layer of the vomeronasal organ. Mice lacking V1RB2 show a disrupted pattern of axonal projections to the accessory olfactory bulb. Mice lacking all but one V1RA and V1RB gene (12% of the V1R repertoire) show a lack of chemosensory response to a subset of known pheromonal ligands and changes in maternal aggression as well as male reproductive behavior.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Vomeronasal 1 Receptor B2/B4
Polyclonal
Vmn1r49
Vmn1r44, Vmn1r49, VN1B4, vomeronasal type-1 receptor B4, vomeronasal type-1 receptor A10, vomeronasal type-1 receptor B11, V1ra10, V1rb11, V1rb4, VRi2, V1rb2, VN1B2
Rabbit
100 μL
Primary
Mouse
Antibody
IgG
Immunohistochemistry, Western Blot
Unconjugated
Q9EQ47, Q9WUF1
VMN1R44, VMN1R49
A synthetic peptide from mouse vomeronasal type-1 receptors B2 and B4 conjugated to an immunogenic carrier protein
RUO
113854, 24112
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Lyophilized
Product Suggestions
Product Suggestions