Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Von Willebrand Factor Mouse anti-Human, Alexa Fluor™ 350, Clone: MM0601-5F22, Novus Biologicals™

Mouse Monoclonal Antibody

Manufacturer:  Novus Biologicals NBP212443AF350

Catalog No. NP212443A30


Add to cart

Description

Description

Von Willebrand Factor Monoclonal antibody specifically detects Von Willebrand Factor in Human samples. It is validated for Western Blot.
Specifications

Specifications

Von Willebrand Factor
MM0601-5F22
Western Blot
Conjugated, Purified
VWF
Human
Protein G purified
Store at 4°C in the dark.
Monoclonal
This antibody is specific to the A2 domain of Von Willebrand Factor
Western Blot
Alexa Fluor™ 350
50 mM sodium borate with 0.05% sodium azide
coagulation factor VIII VWF, F8, F8VWF, von Willebrand factor, VWD, vWF
Mouse
IgG1
0.1 mL
Primary
7450.0
Human
SDS
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit