Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Von Willebrand Factor Mouse anti-Human, Alexa Fluor™ 532, Clone: 50000000, Novus Biologicals™

Mouse Monoclonal Antibody

Manufacturer:  Novus BiologicalsSupplier Diversity Partner NBP223538AF532

Catalog No. NP22338A32


Add to cart

Description

Description

Von Willebrand Factor Monoclonal antibody specifically detects Von Willebrand Factor in Human samples. It is validated for ELISA.
Specifications

Specifications

Von Willebrand Factor
50000000
ELISA
coagulation factor VIII VWF, F8, F8VWF, von Willebrand factor, VWD, vWF
Mouse
IgG1 Kappa
Store at 4°C in the dark.
Monoclonal
Clone 5E7 is specific for human vWf from human serum or plasma
ELISA
Alexa Fluor™ 532
50 mM sodium borate with 0.05% sodium azide
VWF
Factor VIII complex isolated from human plasma adsorbed onto aluminum hydroxide gel
0.1 mL
Primary
7450
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit