Learn More
Von Willebrand Factor Mouse anti-Human, Alexa Fluor™ 594, Clone: MM0601-5F22, Novus Biologicals™
Mouse Monoclonal Antibody
Manufacturer:  Novus BiologicalsSupplier Diversity Partner NBP212443AF594
Catalog No. NP212443A94
Add to cart
Description
Description
Von Willebrand Factor Monoclonal antibody specifically detects Von Willebrand Factor in Human samples. It is validated for Western Blot.
Specifications
Specifications
Von Willebrand Factor
MM0601-5F22
Western Blot
coagulation factor VIII VWF, F8, F8VWF, von Willebrand factor, VWD, vWF
Mouse
IgG1
Store at 4°C in the dark.
Monoclonal
This antibody is specific to the A2 domain of Von Willebrand Factor
Western Blot
Alexa Fluor™ 594
50 mM sodium borate with 0.05% sodium azide
VWF
Human
0.1 mL
Primary
7450
Human