Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Von Willebrand Factor Mouse anti-Human, Clone: rVWF/2480, Novus Biologicals™

Mouse Monoclonal Antibody

$159.82 - $374.34

Specifications

Antigen Von Willebrand Factor
Clone rVWF/2480
Concentration 0.2 mg/ml
Dilution Immunohistochemistry-Paraffin 1-2 ug/ml
Applications Immunohistochemistry (Paraffin)
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
NB121873
View Documents
Novus Biologicals
NBP30720010UG
100 μg
Each for $374.34
Add to cart
 
NB121874
View Documents
Novus Biologicals
NBP30720020UG
20 μg
Each for $159.82
Add to cart
 
Description

Description

Von Willebrand Factor Monoclonal antibody specifically detects Von Willebrand Factor in Human samples. It is validated for Immunohistochemistry (Paraffin)
Specifications

Specifications

Von Willebrand Factor
0.2 mg/ml
Immunohistochemistry (Paraffin)
Purified
RUO
10 mM PBS with 0.05% BSA
7450
IgG1 Kappa
Primary
Store at 4°C.
rVWF/2480
Immunohistochemistry-Paraffin 1-2 ug/ml
Unconjugated
Mouse
Human
coagulation factor VIII VWF, F8, F8VWF, von Willebrand factor, VWD, vWF
Recombinant full-length human Von Willebrand Factor protein
Monoclonal
Protein A or G purified
SDS
Product Certifications
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit