Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Von Willebrand Factor Rabbit anti-Human, Clone: VWF/1859R, Novus Biologicals™

Rabbit Monoclonal Antibody

$159.82 - $406.52

Specifications

Antigen Von Willebrand Factor
Clone VWF/1859R
Concentration 0.2 mg/ml
Dilution Western Blot 1-2 ug/ml, Immunohistochemistry-Paraffin 1-2 ug/ml
Applications Western Blot, Immunohistochemistry (Paraffin)
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
NB121908
View Documents
Novus Biologicals
NBP30721720UG
20 μg
Each for $159.82
Add to cart
 
NB121907
View Documents
Novus Biologicals
NBP30721710UG
100 μg
Each for $406.52
Add to cart
 
Description

Description

Von Willebrand Factor Monoclonal antibody specifically detects Von Willebrand Factor in Human samples. It is validated for Western Blot, Immunohistochemistry (Paraffin)
Specifications

Specifications

Von Willebrand Factor
0.2 mg/ml
Western Blot, Immunohistochemistry (Paraffin)
Purified
RUO
10 mM PBS with 0.05% BSA
7450
IgG
Primary
Store at 4°C.
VWF/1859R
Western Blot 1-2 ug/ml, Immunohistochemistry-Paraffin 1-2 ug/ml
Unconjugated
Rabbit
Human
coagulation factor VIII VWF, F8, F8VWF, von Willebrand factor, VWD, vWF
Recombinant fragment of human Von Willebrand Factor protein (aa1815-1939) (exact sequence is proprietary)
Monoclonal
Protein A purified
SDS
Product Certifications
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit