Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

von Willebrand factor (vWF) Mouse anti-Human, Clone: AvW3, MilliporeSigma™

Mouse Monoclonal Antibody

$145.80 - $373.00

Specifications

Antigen von Willebrand factor (vWF)
Clone AvW3
Concentration 1.0 mg/mL
Applications ELISA, Functional Assay, Immunoprecipitation
Conjugate Unconjugated
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
MABS2151100
View Documents
MilliporeSigma
MABS2151100UG
100 μg
Each for $373.00
Add to cart
 
MABS215125
View Documents
MilliporeSigma
MABS215125UG
25 μg
Each for $145.80
Add to cart
 
Description

Description

von Willebrand factor (vWF) Monoclonal antibody specifically detects von Willebrand factor (vWF) in Human samples. It is validated for ELISA, Inhibits Activity/Function, Immunoprecipitation.
Specifications

Specifications

von Willebrand factor (vWF)
1.0 mg/mL
Unconjugated
Mouse
Human
Purified human vWF antigen II (aa 23-763).
Primary
Stable for 1 year at +2°C to +8°C from date of receipt.
AvW3
ELISA, Functional Assay, Immunoprecipitation
Purified
RUO
Purified mouse monoclonal antibody IgG1 in PBS without preservatives.
Monoclonal
Protein G purified
SDS
Product Certifications
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit