Learn More
VPS11 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech Non-distribution product as customer accommodation.
Rabbit Polyclonal Antibody
Manufacturer:  Proteintech Group IncSupplier Diversity Partner 191401AP
Catalog No. 50-173-2354
Add to cart
Description
Description
Vesicle mediated protein sorting plays an important role in segregation of intracellular molecules into distinct organelles. Genetic studies in yeast have identified more than 40 vacuolar protein sorting (VPS) genes involved in vesicle transport to vacuoles. This gene encodes the human homolog of yeast class C Vps11 protein. The mammalian class C Vps proteins are predomitly associated with late endosomes/lysosomes, and like their yeast counterparts, may mediate vesicle trafficking steps in the endosome/lysosome pathway. Alternative splicing results in multiple transcript variants.
Specifications
Specifications
VPS11
0.17 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Q91W86, Q9H270
END1, hVPS11, PEP5, RING finger protein 108, RNF108, VPS11
Rabbit
IgG
150 μL
-20°C
Primary
315600, 55823, 71732
Flow Cytometry, Immunocytochemistry, Immunofluorescence, Immunoprecipitation, Western Blot
Unconjugated
VPS11
Liquid
Vps11
VPS11 Fusion Protein Ag6227
Antigen Affinity Chromatography
RUO
Antibody
Polyclonal
Human, Mouse, Rat