Learn More
VPS24 Rabbit anti-Human, Polyclonal, Proteintech Non-distribution product as customer accommodation.
Rabbit Polyclonal Antibody
Manufacturer:  Proteintech Group IncSupplier Diversity Partner 154721AP
Catalog No. 50-173-0255
Add to cart
Description
Description
This gene encodes a protein with a conserved RNA-binding motif. The protein is found predomitly in the nucleus, although it is also present in the cytoplasm. It is preferentially associated with mRNAs produced by splicing, including both nuclear mRNAs and newly exported cytoplasmic mRNAs. It is thought that the protein remains associated with spliced mRNAs as a tag to indicate where introns had been present, thus coupling pre- and post-mRNA splicing events. Previously, it was thought that two genes encode this protein, RBM8A and RBM8B; it is now thought that the RBM8B locus is a pseudogene. Two alternative start codons result in two forms of the protein, and this gene also uses multiple polyadenylation sites.
Specifications
Specifications
VPS24
0.27 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Q9Y3E7
CGI 149, CHMP3, Chromatin modifying protein 3, hVps24, NEDF, VPS24
Rabbit
IgG
150 μL
-20°C
Primary
51652
Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot
Unconjugated
CHMP3
Liquid
CHMP3
VPS24 Fusion Protein Ag7776
Antigen Affinity Chromatography
RUO
Antibody
Polyclonal
Human