Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ VPS25 Polyclonal Antibody
Goat Polyclonal Antibody
Supplier: Invitrogen™ PA518464
Description
This antibody is predicted to react with canine, mouse and rat based on sequence homology. This antibody is tested in Peptide ELISA: antibody detection limit dilution 32,000.
VPS25 (Vacuolar Protein Sorting 25 Homolog) is a Protein Coding gene. Among its related pathways are Delta508-CFTR traffic / ER-to-Golgi in CF and Budding and maturation of HIV virion. Gene Ontology (GO) annotations related to this gene include protein homodimerization activity and protein N-terminus binding.
Specifications
VPS25 | |
Polyclonal | |
Unconjugated | |
VPS25 | |
1110020N13Rik; AW107467; D11Wsu68e; dermal papilla-derived protein 9; DERP9; Eap20; ELL-associated protein of 20 kDa; ESCRT-II complex subunit VPS25; FAP20; HGNC:28122; hVps25; MGC10540; vacuolar protein sorting 25 (yeast); vacuolar protein sorting 25 homolog; vacuolar protein sorting 25 homolog (S. cerevisiae); vacuolar protein-sorting-associated protein 25; VPS25 | |
Goat | |
Ammonium sulfate precipitation | |
RUO | |
681059, 84313 | |
-20°C, Avoid Freeze/Thaw Cycles | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot | |
0.5 mg/mL | |
TBS with 0.5% BSA and 0.02% sodium azide; pH 7.3 | |
P0C0A1, Q9BRG1 | |
VPS25 | |
Synthetic peptide sequence (QPNVDTRQKQ) corresponding to the internal amino acids of VPS25 (aa 20-29). | |
100 μg | |
Primary | |
Human, Rat | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction