Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

VPS26A Rabbit anti-Human, Mouse, Rat, Clone: JB93-41, Novus Biologicals™
SDP

Rabbit Monoclonal Antibody

Supplier:  Novus Biologicals NBP275709

Catalog No. NBP275709


Add to Cart

Description

Description

VPS26A Monoclonal specifically detects VPS26A in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunoprecipitation, Immunohistochemistry-Paraffin.
Specifications

Specifications

VPS26A
Monoclonal
1 mg/mL
Western Blot 1:100-1:2000, Immunohistochemistry 1:50-1:200, Immunoprecipitation 1:10 - 1:500, Immunohistochemistry-Paraffin 1:50-1:200
FLJ12930, HB58, Hbeta58, hVPS26, PEP8A, vacuolar protein sorting 26 (yeast homolog), vacuolar protein sorting 26 homolog A (S. pombe), vacuolar protein sorting 26 homolog A (yeast), vacuolar protein sorting-associated protein 26A, Vesicle protein sorting 26A, VPS26vacuolar protein sorting 26 (yeast)
Rabbit
38 kDa
100 μL
Primary
Human, Mouse, Rat
Purified
Western Blot, Immunohistochemistry, Immunoprecipitation, Immunohistochemistry (Paraffin)
JB93-41
Unconjugated
TBS (pH7.4), 0.05% BSA, 40% Glycerol with 0.05% Sodium Azide
VPS26A
Recombinant protein within Human VPS26A aa 210-327/ 327. (SwissProt: O75436 Human; SwissProt: P40336 Mouse; SwissProt: Q6AY86 Rat)
Protein A purified
Autophagy
9559.0
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
IgG
Product Suggestions

Product Suggestions

SDS
Documents

Documents

Product Certifications
Promotions

Promotions

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit