Learn More
VPS26B Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
SDP
Rabbit Polyclonal Antibody
Manufacturer:  Proteintech Group Inc 159151AP150UL
Encompass_Preferred
 View more versions of this product
Catalog No. 50-173-0572
Add to cart
Description
Description
In mammals, there are two paralogues of yeast Vps26, VPS26A and VPS26B. VPS26 is a component of the retromer complex composed of VPS26 (VPS26A or VPS26B), VPS29, VPS35, SNX1 and SNX2. VPS26A and VPS26B subunits define distinct retromer complexes. The retromer complex is important in recycling transmembrane receptors from endosomes to the trans-Golgi network (TGN).
Specifications
Specifications
VPS26B
Polyclonal
Unconjugated
VPS26B
Pep8b, Vesicle protein sorting 26B, VPS26B
Rabbit
Antigen Affinity Chromatography
RUO
112936, 300472, 69091
-20°C
Liquid
Western Blot
0.13 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Q4G0F5, Q8C0E2
Vps26b
VPS26B Fusion Protein Ag8708
150 μL
Primary
Human, Mouse, Rat
Antibody
IgG