Learn More
VPS33A Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech Non-distribution product as customer accommodation.
Rabbit Polyclonal Antibody
Manufacturer:  Proteintech Group IncSupplier Diversity Partner 168961AP
Catalog No. 50-173-1243
Add to cart
Description
Description
Vesicle mediated protein sorting plays an important role in segregation of intracellular molecules into distinct organelles. Genetic studies in yeast have identified more than 40 vacuolar protein sorting (VPS) genes involved in vesicle transport to vacuoles. This gene is a member of the Sec-1 domain family, and it encodes a protein similar to the yeast class C Vps33 protein. The mammalian class C VPS proteins are predomitly associated with late endosomes/lysosomes, and like their yeast counterparts, may mediate vesicle trafficking steps in the endosome/lysosome pathway.
Specifications
Specifications
VPS33A
0.18 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Q63615, Q96AX1, Q9D2N9
hVPS33A, VPS33A
Rabbit
IgG
150 μL
-20°C
Primary
65081, 65082, 77573
Immunoprecipitation, Western Blot
Unconjugated
VPS33A
Liquid
Vps33a
VPS33A Fusion Protein Ag10422
Antigen Affinity Chromatography
RUO
Antibody
Polyclonal
Human, Mouse, Rat