Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
VPS37D Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Supplier: Thermo Scientific PA551162
Description
The antibody detects endogenous levels of total VPS37D protein.
VPS37D Polyclonal antibody specifically detects VPS37D in Human samples. It is validated for Immunohistochemistry (Paraffin).Specifications
VPS37D | |
Polyclonal | |
Unconjugated | |
VPS37D | |
ESCRT-I complex subunit VPS37D; vacuolar protein sorting 37 homolog D; vacuolar protein sorting 37D; vacuolar protein sorting-associated protein 37D; VPS37D; VPS37D, ESCRT-I subunit; WBSCR24; Williams Beuren syndrome chromosome region 24; williams-Beuren syndrome chromosomal region 24 protein; Williams-Beuren syndrome critical region protein 24; williams-Beuren syndrome region protein 24 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
155382 | |
-20°C | |
Liquid |
Immunohistochemistry (Paraffin) | |
1.7 mg/mL | |
PBS with 40% glycerol and 0.05% sodium azide; pH 7.4 | |
Q86XT2 | |
VPS37D | |
Fusion protein of human VPS37D | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction