Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

VRK1 Mouse anti-Human, HRP, Clone: OTI2C9, Novus Biologicals™

Mouse Monoclonal Antibody

Manufacturer:  Novus Biologicals NBP274855H

Catalog No. NBP274855H


Add to cart

Description

Description

VRK1 Monoclonal antibody specifically detects VRK1 in Human samples. It is validated for Western Blot.
Specifications

Specifications

VRK1
OTI2C9
Western Blot
Affinity Purified, Conjugated
VRK1
Recombinant protein fragment corresponding to amino acids 1-230 of human VRK1 (NP_003375) produced in E.coli.
Immunogen affinity purified
Core ESC Like Genes, Protein Kinase, Stem Cell Markers
Primary
7443.0
Western Blot
HRP
PBS with No Preservative
EC 2.7.11.1, MGC117401, MGC138280, MGC142070, PCH1, serine/threonine-protein kinase VRK1, vaccinia related kinase 1, vaccinia virus B1R-related kinase 1, Vaccinia-related kinase 1
Mouse
IgG1
0.1 mL
Store at 4°C in the dark.
Monoclonal
Human
SDS
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit