Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
VRK1 Antibody, Alexa Fluor™ 750, Novus Biologicals™
Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP299819AF750
Description
VRK1 Polyclonal specifically detects VRK1 in Human samples. It is validated for Western Blot, ELISA.Specifications
VRK1 | |
Polyclonal | |
Western Blot, ELISA | |
EC 2.7.11.1, MGC117401, MGC138280, MGC142070, PCH1, serine/threonine-protein kinase VRK1, vaccinia related kinase 1, vaccinia virus B1R-related kinase 1, Vaccinia-related kinase 1 | |
Produced in rabbits immunized with purified, recombinant VRK1 (Q99986; Met1-Lys396). | |
0.1 mL | |
Core ESC Like Genes, Protein Kinase, Stem Cell Markers | |
7443 | |
Store at 4°C in the dark. | |
IgG |
Western Blot, ELISA | |
Alexa Fluor 750 | |
50mM Sodium Borate | |
Rabbit | |
Affinity purified | |
RUO | |
Primary | |
Human | |
Purified |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction