Learn More
VSIG1 Rabbit anti-Human, Alexa Fluor™ 700, Clone: 1314D, R&D Systems™
Rabbit Monoclonal Antibody
Manufacturer:  R&D Systems FAB48181N100UG
Catalog No. FB48181N100
Add to cart
This item is not returnable. View return policy
Description
Description
VSIG1 Monoclonal antibody specifically detects VSIG1 in Human samples. It is validated for Flow Cytometry.
Specifications
Specifications
VSIG1
1314D
Flow Cytometry 0.25-1 ug/10^6 cells
Q86XK7
Cell surface A33 antigen, dJ889N15.1, Glycoprotein A34, GPA34, MGC44287,1700062D20Rik, V-set and immunoglobulin domain containing 1, V-set and immunoglobulin domain-containing protein 1
Mouse myeloma cell line, NS0-derived human VSIG-1, Val22-Gly234, Accession No. Q86XK7
Protein A or G purified from cell culture supernatant
Protect from light. Do not freeze. 12 months from date of receipt, 2 to 8°C as supplied.
Monoclonal
Detects human VSIG-1 in direct ELISAs.
Flow Cytometry
Alexa Fluor 700
Supplied 0.2 mg/mL in a saline solution containing BSA and sodium azide.
Purified, Supernatant, Conjugated
Rabbit
IgG
100 μg
Primary
340547.0
Human