Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human VSIG2 Alexa Fluor 350-conjugated Antibody, R&D Systems™
Mouse Monoclonal Antibody
Supplier: R&D Systems FAB51771U100UG
This item is not returnable.
View return policy
Description
VSIG2 Monoclonal specifically detects VSIG2 in Human samples. It is validated for Flow Cytometry.Specifications
VSIG2 | |
Monoclonal | |
Alexa Fluor 350 | |
Supplied 0.2 mg/mL in a saline solution containing BSA and sodium azide. | |
2210413P10Rik, Cortical thymocyte-like protein, CTH, CTHCTXLcortical thymocyte receptor (X. laevis CTX) like, CT-like protein, CTXL, V-set and immunoglobulin domain containing 2, V-set and immunoglobulin domain-containing protein 2 | |
Mouse myeloma cell line NS0-derived recombinant human VSIG2, Val24-Ala243, Accession No. Q961Q7-1 | |
100 μg | |
23584.0 | |
Human | |
Supernatant |
Flow Cytometry | |
541522 | |
Flow Cytometry 0.25-1 ug/10^6 cells | |
Q961Q7-1 | |
Mouse | |
Protein A or G purified from hybridoma culture supernatant | |
Primary | |
Detects human VSIG2 in direct ELISAs. | |
Protect from light. Do not freeze. 12 months from date of receipt, 2 to 8°C as supplied. | |
IgG1 |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction