Learn More
VSIG2 Mouse anti-Human, Alexa Fluor™ 405, Clone: 541522, R&D Systems™
Mouse Monoclonal Antibody
Manufacturer:  R&D Systems FAB51771V100UG
Catalog No. FB51771V100
Add to cart
This item is not returnable. View return policy
Description
Description
VSIG2 Monoclonal specifically detects VSIG2 in Human samples. It is validated for Flow Cytometry.
Specifications
Specifications
VSIG2
Monoclonal
Alexa Fluor 405
Supplied 0.2 mg/mL in a saline solution containing BSA and sodium azide.
2210413P10Rik, Cortical thymocyte-like protein, CTH, CTHCTXLcortical thymocyte receptor (X. laevis CTX) like, CT-like protein, CTXL, V-set and immunoglobulin domain containing 2, V-set and immunoglobulin domain-containing protein 2
Mouse myeloma cell line NS0-derived recombinant human VSIG2, Val24-Ala243, Accession No. Q961Q7-1
100 μg
23584.0
Human
Supernatant
Flow Cytometry
541522
Flow Cytometry 0.25-1 ug/10^6 cells
Q961Q7-1
Mouse
Protein A or G purified from hybridoma culture supernatant
Primary
Detects human VSIG2 in direct ELISAs.
Protect from light. Do not freeze. 12 months from date of receipt, 2 to 8°C as supplied.
IgG1