Learn More
Novus Biologicals™ VSIG4 Antibody Pair [HRP]
Rabbit Monoclonal Antibody Pair
Manufacturer:  Novus Biologicals™ NBP2793885PLATES
 View more versions of this product
Catalog No. NB009816
Add to cart
Description
Description
This solid phase sandwich ELISA utilizes a monoclonal antibody specific for Human VSIG4.
Specifications
Specifications
Sandwich ELISA
This solid phase sandwich ELISA utilizes a monoclonal antibody specific for Human VSIG4.
31.25 pg/mL
11326
RUO
VSIG4
5 Plates
31.25-2000 pg/mL
CRIg, Ig superfamily protein, Protein Z39Ig, V-set and immunoglobulin domain containing 4, V-set and immunoglobulin domain-containing protein 4, Z39IGcomplement receptor of the immunoglobulin superfamily
HRP
Antibody Pairs
Sandwich ELISA
  • Standard: Each vial contains 100 ng of recombinant VSIG4.
  • Capture Antibody: 1 mg/mL of rabbit anti-VSIG4 monoclonal antibody (in PBS, pH 7.4).
  • Detection Antibody: 0.25 mg/mL rabbit anti-VSIG4 monoclonal antibody conjugated to horseradish-peroxidase (HRP) (in PBS, 50 % HRP-protector, pH 7.4).
  • Storage is content dependent.
    Rabbit
    Human
    VSIG4
    1. Capture Antibody: Dilute to a working concentration of 2 μg/mL in PBS before coating. 2. Detection Antibody: Dilute to working concentration of 0.25 μg/mL in detection antibody dilution buffer before use. 3. Standard: Reconstitute with 1 ml detection antibody dilution buffer. After reconstitution, store at -20 to -80°C in a manual defrost freezer. A seven-point standard curve using 2-fold serial dilutions in sample dilution buffer, and a high standard of 2 ng/mL is recommended.