Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
VSIG4 Mouse anti-Human, Alexa Fluor 488, Clone: 528903, R&D Systems™
Mouse Monoclonal Antibody
Supplier: R&D Systems FAB4646G100UG
This item is not returnable.
View return policy
Description
VSIG4 Monoclonal antibody specifically detects VSIG4 in Human samples. It is validated for Flow CytometrySpecifications
VSIG4 | |
Monoclonal | |
Alexa Fluor 488 | |
Supplied 0.2 mg/mL in a saline solution containing BSA and Sodium Azide. | |
CRIg, Ig superfamily protein, Protein Z39Ig, V-set and immunoglobulin domain containing 4, V-set and immunoglobulin domain-containing protein 4, Z39IG, Z39IGcomplement receptor of the immunoglobulin superfamily | |
Mouse myeloma cell line NS0-derived human VSIG4 protein, Arg20-Pro283, Accession # Q9Y279 | |
100 μg | |
Primary | |
Reconstitution default not known | |
Protect from light. Do not freeze. 12 months from date of receipt, 2 to 8°C as supplied. | |
IgG2a |
Flow Cytometry | |
528903 | |
Flow Cytometry 0.25-1 ug/106 cells | |
Q9Y279 | |
Mouse | |
Protein A or G purified from hybridoma culture supernatant | |
RUO | |
11326 | |
Human | |
Purified |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title