Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

VSIG4 Mouse anti-Human, Clone: 528903, R&D Systems™

Mouse Monoclonal Antibody

$122.30 - $314.00

Specifications

Antigen VSIG4
Clone 528903
Dilution Western Blot 2 ug/mL, Flow Cytometry 0.25 ug/106 cells, CyTOF-ready
Applications Western Blot, Flow Cytometry, CyTOF
Conjugate Unconjugated
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
MAB4646100
View Documents
R&D Systems
MAB4646100
100 μg
Each for $314.00
Add to cart
 
MAB4646SP
View Documents
R&D Systems
MAB4646SP
25 μg
Each for $122.30
Add to cart
 
Description

Description

VSIG4 Monoclonal antibody specifically detects VSIG4 in Human samples. It is validated for Western Blot, Flow Cytometry, CyTOF
Specifications

Specifications

VSIG4
Western Blot 2 ug/mL, Flow Cytometry 0.25 ug/106 cells, CyTOF-ready
Unconjugated
Mouse
Human
Q9Y279
11326
IgG2a
Primary
Reconstitute at 0.5 mg/mL in sterile PBS.
528903
Western Blot, Flow Cytometry, CyTOF
Purified
RUO
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose.
CRIg, Ig superfamily protein, Protein Z39Ig, V-set and immunoglobulin domain containing 4, V-set and immunoglobulin domain-containing protein 4, Z39IG, Z39IGcomplement receptor of the immunoglobulin superfamily
Mouse myeloma cell line NS0-derived human VSIG4 protein, Arg20-Pro283, Accession # Q9Y279
Monoclonal
Protein A or G purified from hybridoma culture supernatant
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70°C as supplied. 1 month, 2 to 8°C under sterile conditions after reconstitution. 6 months, -20 to -70°C under sterile conditions after reconstitution.
SDS
Product Certifications
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit