Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
VSIG4 Rabbit anti-Mouse, Alexa Fluor 405, Polyclonal, Novus Biologicals™
Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP298230AF405
Description
VSIG4 Polyclonal specifically detects VSIG4 in Mouse samples. It is validated for ELISA.Specifications
VSIG4 | |
Polyclonal | |
50 mM Sodium Borate | |
Rabbit | |
Affinity purified | |
RUO | |
11326 | |
Store at 4°C in the dark. | |
IgG |
ELISA | |
Alexa Fluor 405 | |
CRIg, Ig superfamily protein, Protein Z39Ig, V-set and immunoglobulin domain containing 4, V-set and immunoglobulin domain-containing protein 4, Z39IGcomplement receptor of the immunoglobulin superfamily | |
Produced in rabbits immunized with purified, recombinant VSIG4 (NP_808457.1; Met1-Pro187). | |
0.1 mL | |
Primary | |
Mouse | |
Purified |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title