Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

VSIG4 Rat anti-Mouse, PE, Clone: NLA14, eBioscience™ Non-distribution product as customer accommodation.

Rat Monoclonal Antibody

$136.00 - $335.00

Specifications

Classification Monoclonal
Monoclonal or Polyclonal Monoclonal
Primary or Secondary Primary
Antigen VSIG4
Conjugate PE
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
12-575-280 Non-distribution product as customer accommodation.
View Documents
eBioscience
12575280
25 μg
Each for $136.00
Add to cart
 
12-575-282 Non-distribution product as customer accommodation.
View Documents
eBioscience
12575282
100 μg
Each for $335.00
Add to cart
 
Description

Description

This NLA14 monoclonal antibody recognizes mouse V-Set and Immunoglobulin domain containing 4 (VSIG4), also known as Complement Receptor of the Immunoglobulin superfamily (CRIg) or Z39Ig. VSIG4 is a type I transmembrane glycoprotein structurally related to

Applications Reported: This NLA14 antibody has been reported for use in flow cytometric analysis. Applications Tested: This NLA14 antibody has been tested by flow cytometric analysis of resident peritoneal macrophages. This may be used at less than or equal to 0.5 μg per test. A test is defined as the amount (μg) of antibody that will stain a cell sample in a final volume of 100 μL. Cell number should be determined empirically but can range from 10^5 to 10^8 cells/test. It is recommended that the antibody be carefully titrated for optimal performance in the assay of interest. Excitation: 488-561 nm; Emission: 578 nm; Laser: Blue Laser, Green Laser, Yellow-Green Laser T cell activation by APCs is positively and negatively regulated by members of the B7 family. VSIG4 is a strong negative regulator of murine and human T cell proliferation and IL-2 production.
Specifications

Specifications

Monoclonal
Primary
PE
Monoclonal
VSIG4
SDS
Product Certifications
Provide Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit