Learn More
VSIG8 Mouse anti-Human, Alexa Fluor 594, Clone: 961823, R&D Systems™
Mouse Monoclonal Antibody
Manufacturer:  R&D Systems FAB9418T100UG
Catalog No. FAB9418T100
Add to cart
This item is not returnable. View return policy
Description
Description
Specifications
Specifications
VSIG8
961823
Flow Cytometry 0.25-1 ug/10^6 cells
Q5VU13
V-set and immunoglobulin domain containing 8
Human embryonic kidney cell line HEK293-derived transfected with human VSIG8 Met1-Gly263 Accession # Q5VU13
Protein A or G purified from hybridoma culture supernatant
Protect from light. Do not freeze. 12 months from date of receipt, 2 to 8 degreesC as supplied.
Monoclonal
Detects human VSIG8 in direct ELISAs. Stains human VSIG8 transfectants but not irrelevant transfectants in flow cytometry.
Flow Cytometry
Alexa Fluor 594
Supplied 0.2 mg/mL in a saline solution containing BSA and Sodium Azide.
Supernatant
Mouse
IgG1
100 ug
Primary
391123
Human