Learn More
VSNL1 Mouse anti-Human, Clone: OTI7E11, lyophilized, TrueMAB™ Non-distribution product as customer accommodation.
Mouse Monoclonal Antibody
Manufacturer:  OriGeneSupplier Diversity Partner CF802089
Catalog No. 50-166-5859
Add to cart
Description
Description
Reconstitute with PBS (pH 7.3) and recommend to perform another round of desalting process using Product No. 7KMWCO
This gene is a member of the visinin/recoverin subfamily of neuronal calcium sensor proteins. The encoded protein is strongly expressed in granule cells of the cerebellum where it associates with membranes in a calcium-dependent manner and modulates intracellular signaling pathways of the central nervous system by directly or indirectly regulating the activity of adenylyl cyclase. Alternatively spliced transcript variants have been observed, but their full-length nature has not been determined.
Specifications
Specifications
VSNL1
Monoclonal
Unconjugated
VSNL1
Lyophilized
VSNL1
Human recombit protein fragment corresponding to amino acids 2-191 of human VSNL1 produced in E.coli.
Affinity Chromatography
RUO
Antibody
Monoclonal
Human
Immunohistochemistry (Paraffin), Western Blot
OTI7E11
PBS with 8% trehalose and no preservative; pH 7.3
P62760
HLP3, HPCAL3, HUVISL1, VILIP, VILIP-1
Mouse
IgG1
100 μg
-20° C, Avoid Freeze/Thaw Cycles
Primary
7447