Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ VSTM1 Monoclonal Antibody (01)
Mouse Monoclonal Antibody
Supplier: Invitrogen™ MA529881
Description
This product is preservative free. It is recommended to add sodium azide to avoid contamination (final concentration 0.05%-0.1%). This antibody has specificity for Human VSTM1.
VSTM1 (V-Set And Transmembrane Domain Containing 1) is a Protein Coding gene. Gene Ontology (GO) annotations related to this gene include cytokine activity. An important paralog of this gene is LILRB1.
Specifications
VSTM1 | |
Monoclonal | |
1 mg/mL | |
PBS with no preservative | |
Q6UX27 | |
VSTM1 | |
Recombinant Human VSTM1 Protein (Met1-Thr135). | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG1 |
ELISA | |
01 | |
Unconjugated | |
VSTM1 | |
LAIR homolog; OSCAR-like transcript-1; signal inhibitory receptor on leukocytes-1; SIRL1; SIRL-1; UNQ3033; UNQ3033/PRO9835; V-set and transmembrane domain containing 1; V-set and transmembrane domain-containing protein 1; VSTM1 | |
Mouse | |
Protein A | |
RUO | |
284415 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction