Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
VSV-G Epitope Tag Antibody, HRP, Novus Biologicals™
Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NB1002485H
Description
VSV-G Epitope Tag Polyclonal antibody specifically detects VSV-G Epitope Tag in Epitope Tag samples. It is validated for Western Blot, ELISASpecifications
VSV-G Epitope Tag | |
Polyclonal | |
PBS | |
Rabbit | |
Immunogen affinity purified | |
RUO | |
Primary | |
Purified | |
Epitope Tag |
Western Blot, ELISA | |
HRP | |
Vesicular stomatitis virus glycoprotein tag, VSV epitope tag, VSV tag, VSVG, VSV-G epitope tag, vsvg tag | |
Synthetic peptide: YTDIEMNRLGK conjugated to KLH. | |
0.1 mL | |
Epitope Tags | |
Store at 4°C in the dark. | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Product Content Correction