Learn More
VSV-G-tag Rabbit, Polyclonal, GenScript™
Rabbit Polyclonal Antibody
Supplier: Genscript Corporation A0019940

Description
Rabbit Anti-VSV-G-tag Polyclonal Antibody is supplied as a 40 μg aliquot at a concentration of 1 mg/mL in PBS, pH 7.4, containing 0.02% sodium azide. It is purified by immunoaffinity chromatography.
VSV-G-tag Polyclonal antibody specifically detects VSV-G-tag in Tag samples. It is validated for ELISA, Immunoprecipitation, Western BlotSpecifications
VSV-G-tag | |
Polyclonal | |
Unconjugated | |
vesicular stomatitis virus; VSIVgp3; VSV epitope tag; VSV tag; vsvg; VSVG epitope tag; VSV-G epitope tag; VSVG tag; YTDIEMNRLGK | |
VSV-G epitope tag peptide YTDIEMNRLGK conjugated to KLH | |
40 μg | |
Primary | |
Antibody | |
IgG |
ELISA, Immunoprecipitation, Western Blot | |
1 mg/mL | |
PBS with 0.02% sodium azide; pH 7.4 | |
Rabbit | |
Affinity chromatography | |
RUO | |
-20°C or -80°C if preferred | |
Liquid | |
Tag |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.