Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

VSV-Glycoprotein Mouse anti-Virus, Clone: 1E9F9, MilliporeSigma™

Mouse Monoclonal Antibody

$153.40 - $387.00

Specifications

Antigen VSV-Glycoprotein
Clone 1E9F9
Applications ELISA, Flow Cytometry, Immunofluorescence, Immunoprecipitation, Neutralization, Radioimmune Assays, Western Blot
Conjugate Unconjugated
Format Purified
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
MABF2321100
View Documents
MilliporeSigma
MABF2321100UL
100 μL
Each for $387.00
Add to cart
 
MABF232125
View Documents
MilliporeSigma
MABF232125UL
25 μL
Each for $153.40
Add to cart
 
Description

Description

VSV-Glycoprotein Monoclonal antibody specifically detects VSV-Glycoprotein in Virus samples. It is validated for ELISA, Flow Cytometry, Immunofluorescence, Immunoprecipitation, Neutralizing, Radioimmune Assays, Western Blotting.
Specifications

Specifications

VSV-Glycoprotein
ELISA, Flow Cytometry, Immunofluorescence, Immunoprecipitation, Neutralization, Radioimmune Assays, Western Blot
Purified
RUO
Purified mouse monoclonal antibody IgG2a in PBS without preservatives.
IgG2aκ
Primary
Stable for 1 year at -20°C from date of receipt. Handling Recommendations: Upon receipt and prior to removing the cap, centrifuge the vial and gently mix the solution. Aliquot into microcentrifuge tubes and store at -20°C. Avoid repeated freeze/thaw cycles, which may damage IgG and affect product performance.
1E9F9
Unconjugated
Mouse
Virus
Vesicular stomatitis virus, Indiana strain.
Monoclonal
Protein G purified
SDS
Product Certifications
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit