Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

VSV Nucleoprotein Mouse anti-Virus, Clone: 10G4, MilliporeSigma™

Mouse Monoclonal Antibody

$153.40 - $387.00

Specifications

Antigen VSV Nucleoprotein
Clone 10G4
Applications Immunocytochemistry, Immunoprecipitation, Western Blot
Conjugate Unconjugated
Format Purified
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
MABF2348100
View Documents
MilliporeSigma
MABF2348100UG
100 μg
Each for $387.00
Add to cart
 
MABF234825
View Documents
MilliporeSigma
MABF234825UG
25 μg
Each for $153.40
Add to cart
 
Description

Description

VSV Nucleoprotein Monoclonal antibody specifically detects VSV Nucleoprotein in Virus samples. It is validated for Immunocytochemistry, Immunoprecipitation, Western Blotting.
Specifications

Specifications

VSV Nucleoprotein
Immunocytochemistry, Immunoprecipitation, Western Blot
Purified
RUO
Purified mouse monoclonal antibody IgG2a in buffer containing 0.1 M Tris-Glycine (pH 7.4), 150 mM NaCl with 0.05% sodium azide.
IgG2aκ
Primary
Stable for 1 year at 2-8°C from date of receipt.
10G4
Unconjugated
Mouse
Virus
Vesicular stomatitis virus, Indiana serotype.
Monoclonal
Protein G purified
SDS
Product Certifications
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit