Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

VSX1 Rabbit anti-Mouse, Polyclonal, Novus Biologicals™

Rabbit Polyclonal Antibody

$106.19 - $352.89

Specifications

Antigen VSX1
Dilution Western Blot 1.0 ug/ml
Applications Western Blot
Conjugate Unconjugated
Format Purified
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
NB125771
View Documents
Novus Biologicals
NBP310435100UL
100 μg
Each for $352.89
Add to cart
 
NB125772
View Documents
Novus Biologicals
NBP31043525UL
25 μg
Each for $106.19
Add to cart
 
Description

Description

VSX1 Polyclonal antibody specifically detects VSX1 in Mouse samples. It is validated for Western Blot
Specifications

Specifications

VSX1
Western Blot
Purified
RUO
PBS buffer, 2% sucrose
30813
Polyclonal
Affinity purified
Western Blot 1.0 ug/ml
Unconjugated
Rabbit
Mouse
CAASDS, KTCN, KTCN1, PPCD, PPD, RINX, VSX1 visual system homeobox 1
The immunogen is a synthetic peptide directed towards the C-terminal region of Mouse VSX1 (NP_473409). Peptide sequence TGMRKPESEDKLAGLWEFDHLKKGANKDEDGPERGPDETTQNPENSLEDV
Primary
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
SDS
Product Certifications
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit