Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

VSX2 Rabbit anti-Human, Polyclonal , Abnova™

Rabbit polyclonal antibody raised against recombinant human VSX2.

Manufacturer:  Abnova Corporation PAB29547

Catalog No. 89-935-100


Add to cart

Description

Description

Sequence: LGLNKEPPSSHPRAALDGLAPGHLLAARSVLSPAGVGGMGLLGPGGLPGFYTQPTFLEVLSDPQSVHLQPLGRASGPLDTSQTASSDSEDVSSSDRKMSKSALNQTKKRKKRRHRTIFTSYQLEELEKAFNEAH
Specifications

Specifications

VSX2
Unconjugated
Immunohistochemistry (1:50-1:200) Western Blot (1:100-1:250) The optimal working dilution should be determined by the end user.
Liquid
VSX2
Recombinant protein corresponding to human VSX2.
Antigen affinity purification
RUO
Primary
338917
Immunohistochemistry (PFA fixed), Western Blot
Rabbit polyclonal antibody raised against recombinant human VSX2.
In PBS, pH 7.2 (40% glycerol, 0.02% sodium azide)
CHX10/HOX10/MCOP2/MCOPCB3/RET1
Rabbit
IgG
100 uL
Store at 4°C. For long term storage store at -20°C.
Aliquot to avoid repeated freezing and thawing.
Polyclonal
Human
SDS
Documents
Product Certifications
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit