Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
VTI1A Mouse anti-Human, Mouse, Rat, Canine, Monkey, DyLight 550, Clone: OTI1F4, Novus Biologicals
Mouse Monoclonal Antibody
Manufacturer: Novus Biologicals NBP274862R
Description
VTI1A Monoclonal antibody specifically detects VTI1A in Human, Mouse, Rat, Canine, Monkey samples. It is validated for Western Blot, Immunohistochemistry.Specifications
VTI1A | |
OTI1F4 | |
Western Blot, Immunohistochemistry | |
Affinity Purified, Conjugated | |
VTI1A | |
Full length recombinant protein of human VTI1A (NP_660207) produced in HEK293T cells. | |
Immunogen affinity purified | |
Store at 4°C in the dark. | |
Monoclonal | |
Canine, Human, Monkey, Mouse, Rat |
Immunohistochemistry, Western Blot | |
DyLight 550 | |
50mM Sodium Borate with 0.05% Sodium Azide | |
MVti1, vesicle transport through interaction with t-SNAREs homolog 1A, vesicle transport through interaction with t-SNAREs homolog 1A (yeast) | |
Mouse | |
IgG2a | |
0.1 mL | |
Primary | |
143187.0 |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title