Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ VTI1A Monoclonal Antibody (OTI1F4)

Mouse Monoclonal Antibody

Supplier:  Invitrogen™ MA526213

Catalog No. PIMA526213


Only null left
Add to Cart

Description

Description

VTI1A Monoclonal Antibody for Western Blot, IHC (P)

V-SNARE that mediates vesicle transport pathways through interactions with t-SNAREs on the target membrane. These interactions are proposed to mediate aspects of the specificity of vesicle trafficking and to promote fusion of the lipid bilayers. Involved in vesicular transport from the late endosomes to the trans-Golgi network. Along with VAMP7, involved in an non- conventional RAB1-dependent traffic route to the cell surface used by KCNIP1 and KCND2. May be involved in increased cytokine secretion associated with cellular senescence.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

VTI1A
Monoclonal
1 mg/mL
PBS with 1% BSA, 50% glycerol and 0.02% sodium azide; pH 7.3
O89116, Q96AJ9, Q9JI51
VTI1A
Full length human recombinant protein of VTI1A produced in HEK293T cell.
100 μL
Primary
Human, Mouse, Rat, Canine, Monkey
Antibody
IgG2a
Immunohistochemistry (Paraffin), Western Blot
OTI1F4
Unconjugated
VTI1A
1110014F16Rik; 1110018K19Rik; 4921537J05Rik; EGK_20072; MMDS3; MVti1; MVti1a; SNARE Vti1a-beta protein; vesicle transport through interaction with t-SNAREs 1 homolog; vesicle transport through interaction with t-SNAREs 1A; vesicle transport through interaction with t-SNAREs homolog 1A; Vesicle transport v-SNARE protein Vti1-like 2; Vti1; VTI1A; Vti1l2; VTI1RP2; Vti1-rp2
Mouse
Affinity chromatography
RUO
100685447, 143187, 53611, 65277
-20°C, Avoid Freeze/Thaw Cycles
Liquid
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Product Certifications
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.