Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ VTI1B Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA581120
Description
This product is preservative free. It is recommended to add sodium azide to avoid contamination (final concentration 0.05%-0.1%). This antibody has specificity for Human VTI1B.
V-SNARE that mediates vesicle transport pathways through interactions with t-SNAREs on the target membrane. These interactions are proposed to mediate aspects of the specificity of vesicle trafficking and to promote fusion of the lipid bilayers. May be concerned with increased secretion of cytokines associated with cellular senescence. Fischer von Mollard G., J. Biol. Chem. 273:2624-2630(1998). Li H.-C., Biochem. Biophys. Res. Commun. 247:70-74(1998).
Specifications
VTI1B | |
Polyclonal | |
Unconjugated | |
Vti1b | |
A novel SNARE; AU015348; GES30; HGNC:17793; MVti1b; SNARE; Soluble N-ethylmaleimide-sensitive factor-attachment protein receptor; vesicle transport through interaction with t-SNAREs 1B; vesicle transport through interaction with t-SNAREs 1B homolog; vesicle transport through interaction with t-SNAREs homolog 1B; Vesicle transport v-SNARE protein Vti1-like 1; vesicle-associated soluble NSF attachment protein receptor; v-SNARE; VTI1; VTI1B; VTI1L; VTI1L1; VTI1-LIKE; vti1-rp1; VTI2 | |
Rabbit | |
Antigen Affinity Chromatography, Protein A | |
RUO | |
10490 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunoprecipitation, Western Blot | |
1 mg/mL | |
PBS with no preservative | |
Q9UEU0 | |
Vti1b | |
Recombinant Human VTI1B. | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction