Learn More
VU 0155041 sodium salt, Tocris
Potent, positive allosteric mGlu4 agonist; Sodium salt of VU 0155041
Manufacturer:  R&D Systems 3311/10
Quantity 10mg
 View more versions of this product
Catalog No. 33-111-0
  Check Availability
Add to cart
Specifications Description & Specifications
Chemical Identifiers
>98%
338.16
VU 0155041 sodium salt
C14H14Cl2NNaO3
cis-2-[[(3, 5-Dichlorophenyl)amino]carbonyl]cyclohexanecarboxylic acid sodium salt
Specifications
10mg
>98%
338.16
VU 0155041 sodium salt
1259372-69-4
C14H14Cl2NNaO3
cis-2-[[(3, 5-Dichlorophenyl)amino]carbonyl]cyclohexanecarboxylic acid sodium salt
Safety and Handling
Recommended Storage : Desiccate at Room Temperature