VU 0155041 sodium salt, Tocris

Potent, positive allosteric mGlu4 agonist; Sodium salt of VU 0155041

Manufacturer: R&D Systems 3311/50

Quantity 50mg

 View more versions of this product

Catalog No. 33-115-0

  Check Availability
Add to Cart

    Chemical Identifiers

    Assay Percent Range >98%
    Molecular Formula C14H14Cl2NNaO3
    Formula Weight 338.16
    Synonym cis-2-[[(3, 5-Dichlorophenyl)amino]carbonyl]cyclohexanecarboxylic acid sodium salt
    Chemical Name or Material VU 0155041 sodium salt

    Specifications

    Formula Weight 338.16
    Quantity 50mg
    Assay Percent Range >98%
    Molecular Formula C14H14Cl2NNaO3
    CAS 1259372-69-4
    Synonym cis-2-[[(3, 5-Dichlorophenyl)amino]carbonyl]cyclohexanecarboxylic acid sodium salt
    Recommended Storage Desiccate at Room Temperature
    Chemical Name or Material VU 0155041 sodium salt

    Desiccate at Room Temperature

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit