Tocris™ VU 0255035

Highly selective muscarinic M1 antagonist

Manufacturer: Tocris™ 3727/10

 View more versions of this product

Catalog No. 37-271-0R

  Check Availability
Add to Cart

Disclaimers:

For Research Use Only


    Specifications

    Inhibitors VU 0255035
    Purity >99
    Quantity 10mg
    Product Type VU 0255035
    Molecular Weight 432.52

    Highly selective muscarinic M1 antagonist (Ki = 14.87nM). Targets the M1 orthosteric site with greater than 75-fold selectivity over the other receptor subtypes. Reduces pilocarpine-induced seizures in mice. Brain penetrant.

    • Chemical Name: N-[3-Oxo-3-[4-(4-pyridinyl)-1-piperazinyl]propyl]-2,1,3-benzothiadiazole-4-sulfonamide
    • Formula: C18H20N6O3S2
    • CAS Number: 1135243-19-4
    • Solubility: Soluble to 20mM in DMSO
    • Purity: >99

  • Description & Specifications

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit