VU 0357121, Tocris™

Positive allosteric modulator of mGlu5

Manufacturer: R&D Systems 4437/10

Quantity 10mg

 View more versions of this product

Catalog No. 44-371-0

  Check Availability
Add to Cart

    Chemical Identifiers

    Assay Percent Range >99%
    Molecular Formula C17H17F2NO2
    Formula Weight 305.32
    Synonym 4-Butoxy-N-(2, 4-difluorophenyl)benzamide
    Chemical Name or Material VU 0357121

    Specifications

    Formula Weight 305.32
    Quantity 10mg
    Assay Percent Range >99%
    Molecular Formula C17H17F2NO2
    CAS 433967-28-3
    Synonym 4-Butoxy-N-(2, 4-difluorophenyl)benzamide
    Recommended Storage Store at +4°C
    Chemical Name or Material VU 0357121

    Store at +4°C

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit