Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tocris™ VU 0361737

Selective positive allosteric modulator at mGlu4

Manufacturer:  Tocris™ 3707/10

 View more versions of this product

Catalog No. 37-071-0R


  Check Availability
Add to cart

Disclaimers:

For Research Use Only


Specifications

Description

Selective positive allosteric modulator at mGlu4 receptors (EC50 values are 240 and 110nM at human and rat receptors respectively). Inactive at mGlu1, mGlu2, mGlu3, mGlu6 and mGlu7 receptors and displays weak activity at mGlu5 and mGlu8 receptors. Brain penetrant.
  • Chemical Name: N-(4-Chloro-3-methoxyphenyl)-2-pyridinecarboxamide
  • Formula: C13H11ClN2O2
  • CAS Number: 1161205-04-4
  • Solubility: Soluble to 100mM in DMSO
  • Purity: >99
Description & Specifications

Specifications

VU 0361737
>99
10mg
VU 0361737
262.69
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit