Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tocris Bioscience™ VU 0409106

Potent and selective negative allosteric modulator at mGlu5

$164.80 - $693.50

Specifications

Chemical Name or Material 3-Fluoro-N-(4-methyl-2-thiazolyl)-5-(5-pyrimidinyloxy)benzamide
Molecular Formula C15H11FN4O2S
CAS 1276617-62-9
Product Type Target: Group I mGluR Modulators
Formula Weight 330.34
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
53-221-0 View Documents Tocris Bioscience™
5322/10
10mg Each for $164.80
Add to cart
 
53-225-0 View Documents Tocris Bioscience™
5322/50
50mg Each for $693.50
Add to cart
 
Description

Description

Potent and selective mGlu5 negative allosteric modulator (IC50 = 49 nM at human mGlu5). Selective for mGlu5 over a panel of GPCRs and ion channels. Exhibits no significant nonspecific binding in rat brain homogenates in vitro. Inhibits marble burying behavior in mice. Brain penetrant.
Specifications

Specifications

3-Fluoro-N-(4-methyl-2-thiazolyl)-5-(5-pyrimidinyloxy)benzamide
1276617-62-9
330.34
C15H11FN4O2S
Target: Group I mGluR Modulators
Safety and Handling

Safety and Handling

Recommended Storage : Store at 4°C

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit